3,5-Difluoro-4-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A303331-25G
A303331-5G
A303331-10G
A303331-250MG
A303331-1G
103814-368EA
27
USD
103814-368
103835-536
76838-218
76838-220
77141-564
77141-566
77305-866
3,5-Difluoro-4-hydroxybenzaldehyde 97%
3,5-Difluoro-4-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄F₂O₂ Boiling Pt: 198.5 °C (760 mm Hg) Melting Pt: 122…124 °C Flash Pt: 73.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06797917 CAS Number: 118276-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...