3,5-Difluoro-4-formylbenzonitrile 95%

Supplier: Ambeed
Warning

A271138-1G A271138-5G A271138-250MG A271138-50MG
103813-036EA 30 USD
103813-036 103852-946 77092-718 77092-720 77276-776 77276-778
3,5-Difluoro-4-formylbenzonitrile 95%
3,5-Difluoro-4-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₂NO
MW: 167.112 g/mol
Boiling Pt: 261.9 °C (760 mm Hg)
Melting Pt: 97…101 °C
Flash Pt: 112.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08457661
CAS Number: 467442-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR