3,5-Difluoro-4-formylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-2,6-difluorobenzaldehyde

A114045-100MG A114045-250MG A114045-1G
103798-336EA 77 USD
103798-336 103824-178 76821-998 76822-000 77310-998
3,5-Difluoro-4-formylbenzoic acid 97%
3,5-Difluoro-4-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Boiling Pt: 319.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09835188
CAS Number: 736990-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR