3,5-Diethyl-4-hydroxybenzaldehyde 96%

Supplier: Ambeed
Warning

A171493-10G A171493-250MG A171493-1G
103806-640EA 125 USD
103806-640 103844-916 77029-042 77029-044 77301-138
3,5-Diethyl-4-hydroxybenzaldehyde 96%
3,5-Diethyl-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13152308
CAS Number: 69574-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR