2-Amino-3,5-dichloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-2-pyridylamine, 3,5-Dichloropyridin-2-amine

A106720-100G A106720-10G A106720-25G A106720-500G A106720-5G
103822-818EA 7 USD
103822-818 77283-500 77283-502 77283-504 77283-506 77309-672
2-Amino-3,5-dichloropyridine 98%
2-Amino-3,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 235.2 °C (760 mm Hg)
Melting Pt: 81…83 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00006313
CAS Number: 4214-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR