3,5-Dichloropyridazine 95%

Supplier: Ambeed
Warning

A379761-1G A379761-25G A379761-5G A379761-100G A379761-10G A379761-250MG
103826-652EA 26 USD
103826-652 103837-688 76841-000 76841-002 76841-004 77278-516 77278-518 77278-520
3,5-Dichloropyridazine 95%
3,5-Dichloropyridazine

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂
Boiling Pt: 280.8 °C (760 mm Hg)
Flash Pt: 150.5 °C
Storage Temperature: Ambient
MDL Number: MFCD10698048
CAS Number: 1837-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR