3,5-Dichloropyrazine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A210120-10G
A210120-250MG
A210120-1G
A210120-25G
A210120-5G
103817-978EA
14
USD
103817-978
103848-670
76975-510
76975-512
77087-038
77087-040
77087-042
3,5-Dichloropyrazine-2-carboxylic acid 97%
3,5-Dichloropyrazine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₂Cl₂N₂O₂ MW: 192.9876 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09909819 CAS Number: 312736-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...