3,5-Dichlorophenylboronic acid 97%

Supplier: Ambeed
Warning

A205014-1G A205014-5G A205014-10G A205014-25G A205014-100G
103817-582EA 5 USD
103817-582 103848-246 77086-444 77086-446 77097-522 77302-924 77612-002
3,5-Dichlorophenylboronic acid 97%
3,5-Dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Boiling Pt: 351.7 °C (760 mm Hg)
Melting Pt: 302…306 °C
Flash Pt: 166.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00051935
CAS Number: 67492-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR