4-Cyano-3,5-dichloropyridine 97%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dichloro-4-pyridinecarbonitrile, 3,5-Dichloropyridine-4-carbonitrile, 3,5-Dichloroisonicotinonitrile

A381184-25G A381184-5G A381184-10G
103826-692EA 8 USD
103826-692 103837-726 76841-040 76841-042 77278-536
4-Cyano-3,5-dichloropyridine 97%
4-Cyano-3,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂
MW: 173 g/mol
Boiling Pt: 266.2 °C (760 mm Hg)
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00104651
CAS Number: 153463-65-1
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR