3,5-Dichloro-4-pyridinecarboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-4-formylpyridine, 3,5-Dichloropyridine-4-carboxaldehyde, 3,5-Dichloroisonicotinaldehyde

A176420-100G A176420-10G A176420-250MG A176420-500G A176420-1G A176420-25G A176420-5G
103807-132EA 6 USD
103807-132 103845-460 76998-374 76998-376 76998-378 76998-380 77082-810 77082-812 77082-814
3,5-Dichloro-4-pyridinecarboxaldehyde 97%
3,5-Dichloro-4-pyridinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176.00 g/mol
Melting Pt: 75…79 °C
Flash Pt: 100.4 °C
Storage Temperature: Ambient
MDL Number: MFCD04039921
CAS Number: 136590-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR