3,5-Dichlorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A183010-1G A183010-25G A183010-5G A183010-100G A183010-10G
103815-592EA 6 USD
103815-592 103846-154 77083-740 77083-742 77083-744 77138-784 77138-786
3,5-Dichlorobenzyl alcohol 98%
3,5-Dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 268.4 °C (760 mm Hg)
Melting Pt: 79…82 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00004634
CAS Number: 60211-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR