3,5-Dichlorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dichlorobenzene-1-sulphonyl chloride

A585537-25G A585537-5G A585537-10G A585537-1G
103840-030EA 15 USD
103840-030 76844-162 76844-164 77157-466 77157-468
3,5-Dichlorobenzenesulfonyl chloride 98%
3,5-Dichlorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Boiling Pt: 158…160 °C (15 mm Hg)
Melting Pt: 30…33 °C
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00051698
CAS Number: 705-21-5
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR