3,5-Dichlorobenzaldehyde 98%

Supplier: Ambeed
Danger

A455403-10G A455403-25G A455403-5G
103838-912EA 10 USD
103838-912 77034-146 77307-388 77307-390
3,5-Dichlorobenzaldehyde 98%
3,5-Dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Melting Pt: 63.5…65.5 °C
Flash Pt: 235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003352
CAS Number: 10203-08-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR