3,5-Dichloro-4-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A180061-100G A180061-100MG A180061-10G A180061-250MG A180061-1G A180061-25G A180061-5G
103807-504EA 12 USD
103807-504 103845-858 76974-424 76974-426 76974-428 76974-430 77083-336 77083-338 77083-340
3,5-Dichloro-4-methylbenzoic acid 98%
3,5-Dichloro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C8H6Cl2O2
MW: 205.0400 g/mol
Boiling Pt: 330.9 °C (760 mm Hg)
Melting Pt: 185…187 °C
Flash Pt: 153.9 °C
Storage Temperature: Ambient
MDL Number: MFCD09743850
CAS Number: 39652-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR