3,5-Dichloro-4-methylaniline 97%

Supplier: Ambeed
Warning

A110559-10G A110559-250MG A110559-1G A110559-5G
103797-684EA 38 USD
103797-684 103823-490 77270-868 77270-870 77310-288 77310-290
3,5-Dichloro-4-methylaniline 97%
3,5-Dichloro-4-methylaniline

Precaución: for Research Use Only

Formula: C7H7Cl2N
Boiling Pt: 284.6 °C (760 mm Hg)
Flash Pt: 125.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00266300
CAS Number: 54730-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR