3,5-Dichloro-4-hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A204560-1G A204560-25G A204560-5G A204560-10G A204560-250MG
103817-560EA 13 USD
103817-560 103848-222 77086-392 77086-394 77086-396 77112-278 77685-520
3,5-Dichloro-4-hydroxybenzaldehyde 98%
3,5-Dichloro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
Boiling Pt: 256.7 °C (760 mm Hg)
Melting Pt: 154 °C
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017605
CAS Number: 2314-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR