3,5-Dichloro-4-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

A146841-100G A146841-10G A146841-250MG A146841-1G A146841-25G A146841-5G
103803-706EA 43 USD
103803-706 103841-800 77272-826 77272-828 77272-830 77299-546 77317-142 77317-144
3,5-Dichloro-4-fluorobenzonitrile 97%
3,5-Dichloro-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂Cl₂FN
MW: 190.0019 g/mol
Boiling Pt: 235.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09835204
CAS Number: 103879-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR