3,5-dichloro-2-nitropyridine 98%

Supplier: Ambeed
Warning

A305618-10G A305618-250MG A305618-1G A305618-5G
103814-440EA 104 USD
103814-440 103835-606 76816-828 76816-830 76838-302 76838-304
3,5-dichloro-2-nitropyridine 98%
3,5-dichloro-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06254585
CAS Number: 610278-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR