3,5-Dichlorosalicylaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde

A204882-25G A204882-5G A204882-10G A204882-500G A204882-100G
103848-240EA 9 USD
103848-240 77086-432 77086-434 77112-290 77112-292 77567-964
3,5-Dichlorosalicylaldehyde 97%
3,5-Dichlorosalicylaldehyde

Precaución: For research use only.

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Melting Pt: 95…97 °C
Flash Pt: 96.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00003320
CAS Number: 90-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR