3,5-Dibromotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dibromo-5-methylbenzene

A125625-100G A125625-10G A125625-5G A125625-25G A125625-500G A125625-1000G
77136-694EA 41.38 USD
77136-694 77136-696 77298-080 77312-850 77614-984 77685-094
3,5-Dibromotoluene 98%
3,5-Dibromotoluene

Precaución: for Research Use Only

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 246 °C (760 mm Hg)
Melting Pt: 37…41 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00013528
CAS Number: 1611-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR