3,5-Dibromophenylboronic acid 98%

Supplier: Ambeed
Warning

A124094-100G A124094-10G A124094-250MG A124094-25G A124094-1G A124094-5G
103800-482EA 6 USD
103800-482 77094-646 77094-648 77094-650 77094-652 77312-612 77312-614
3,5-Dibromophenylboronic acid 98%
3,5-Dibromophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBr₂O₂
MW: 279.72 g/mol
Boiling Pt: 382.8 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 185.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01075725
CAS Number: 117695-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR