1-Chloro-3,5-dibromobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene

A203030-25G A203030-5G A203030-100G A203030-10G A203030-500G
103848-086EA 5 USD
103848-086 77086-200 77086-202 77152-228 77152-230 77152-232
1-Chloro-3,5-dibromobenzene 97%
1-Chloro-3,5-dibromobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂Cl
MW: 270.35
Melting Pt: 91…94 °C
Flash Pt: 115.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00070765
CAS Number: 14862-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR