3,5-Dibromobenzoic acid 97%

Supplier: Ambeed
Warning

A126573-100G A126573-10G A126573-500G A126573-5G A126573-25G
77108-882EA 77.25 USD
77108-882 77108-884 77108-886 77298-138 77313-062
3,5-Dibromobenzoic acid 97%
3,5-Dibromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Br₂O₂
Boiling Pt: 355.2 °C (760 mm Hg)
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00051758
CAS Number: 618-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR