3,5-Dibromobenzene-1-sulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dibromobenzene-1-sulphonyl chloride

A196938-100MG A196938-250MG A196938-1G A196938-5G
103816-878EA 34 USD
103816-878 103847-494 76814-114 76814-116 77085-468 77085-470
3,5-Dibromobenzene-1-sulfonyl chloride 97%
3,5-Dibromobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃Br₂ClO₂S
MW: 334.41 g/mol
Boiling Pt: 365.1 °C (760 mm Hg)
Flash Pt: 175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12025425
CAS Number: 39213-20-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR