3,5-Dibromoaniline 98%

Supplier: Ambeed
Warning

A313519-100G A313519-10G A313519-500G A313519-25G A313519-5G A313519-1G
103825-006EA 5 USD
103825-006 103835-918 76787-968 76787-970 76787-972 76838-736 76838-738 77306-090
3,5-Dibromoaniline 98%
3,5-Dibromoaniline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
Flash Pt: 133.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00047841
CAS Number: 626-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR