3,5-Dibromo-4-nitro-1H-pyrazole 95%

Supplier: Ambeed
Warning

A164062-250MG A164062-1G A164062-25G A164062-5G
103805-806EA 18 USD
103805-806 103844-072 77273-464 77320-200 77320-202 77320-204
3,5-Dibromo-4-nitro-1H-pyrazole 95%
3,5-Dibromo-4-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C3HBr2N3O2
MW: 270.8669 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09972275
CAS Number: 104599-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR