3,5-Dibromo-4-picoline 98%

Supplier: Ambeed
Warning

A132983-10G A132983-250MG A132983-1G A132983-25G A132983-5G A132983-100G
103801-808EA 6 USD
103801-808 77095-030 77095-032 77314-394 77314-396 77314-398 77685-151
3,5-Dibromo-4-picoline 98%
3,5-Dibromo-4-picoline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
Boiling Pt: 243 °C (760 mm Hg)
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00128859
CAS Number: 3430-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR