3,5-Dibromo-4-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A187785-100MG A187785-250MG A187785-1G A187785-5G
103816-042EA 28 USD
103816-042 103846-636 76813-808 76813-810 77084-348 77302-100
3,5-Dibromo-4-methoxybenzaldehyde 97%
3,5-Dibromo-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Boiling Pt: 332.0±37.0 °C (760 mm Hg)
Melting Pt: 90…91 °C
Flash Pt: 154.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00858984
CAS Number: 108940-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR