3,5-Dibromo-4-chloropyridine 98%

Supplier: Ambeed
Warning

A228760-100G A228760-10G A228760-25G A228760-500G A228760-1G A228760-5G A228760-250MG
103819-416EA 12 USD
103819-416 103850-320 76976-058 76976-060 76976-062 76976-064 77089-120 77089-122 77612-224
3,5-Dibromo-4-chloropyridine 98%
3,5-Dibromo-4-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂ClN
Boiling Pt: 256.4±35.0 °C (760 mm Hg)
Melting Pt: 98 °C
Flash Pt: 108.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233993
CAS Number: 13626-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR