3,5-Dibromo-2-nitropyridine 97%

Supplier: Ambeed
Warning

A132847-100MG A132847-250MG A132847-1G A132847-5G
103801-796EA 175 USD
103801-796 77272-080 77272-082 77298-590 77298-592
3,5-Dibromo-2-nitropyridine 97%
3,5-Dibromo-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂N₂O₂
MW: 281.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01612785
CAS Number: 610261-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR