3,5-Dibromo-2-methylpyridine 97%

Supplier: Ambeed
Warning

A173537-100G A173537-10G A173537-25G A173537-500G A173537-1G A173537-5G
103806-826EA 8 USD
103806-826 103845-126 76998-298 76998-300 76998-302 76998-304 77082-372 77082-374
3,5-Dibromo-2-methylpyridine 97%
3,5-Dibromo-2-methylpyridine

Precaución: for Research Use Only

Formula: C6H5Br2N
MW: 250.9186 g/mol
Boiling Pt: 227.9 °C (760 mm Hg)
Flash Pt: 91.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00234159
CAS Number: 38749-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR