3,5-Dibromo-2-pyridone 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-2-hydroxypyridine, 3,5-Dibromo-2-pyridinol

A136524-100G A136524-10G A136524-1G A136524-25G A136524-5G
103802-312EA 13 USD
103802-312 77272-258 77272-260 77315-212 77315-214 77315-216
3,5-Dibromo-2-pyridone 98%
3,5-Dibromo-2-pyridone

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO
MW: 252.89 g/mol
Boiling Pt: 305.4 °C (760 mm Hg)
Melting Pt: 204…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00023472
CAS Number: 13472-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR