3,5-Dibromo-2-hydroxypyrazine 98%
Supplier: Ambeed
|
Warning
|
A639664-1G
A639664-250MG
103840-338EA
26
USD
103840-338
76844-514
77280-466
3,5-Dibromo-2-hydroxypyrazine 98%
3,5-Dibromo-2-hydroxypyrazine
Precaución: for Research Use Only
Formula:
C4H2Br2N2O Boiling Pt: 430.5 °C (760 mm Hg) Melting Pt: 174…178 °C Flash Pt: 214.151 °C Storage Temperature: Ambient |
MDL Number:
MFCD07371406 CAS Number: 21943-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...