3,5-Dibromo-1-methylpyrazin-2(1H)-one 97%

Supplier: Ambeed
Warning

A210036-100G A210036-1G A210036-25G A210036-5G A210036-250MG
103817-976EA 30 USD
103817-976 103848-664 77030-476 77087-030 77087-032 77087-034 77600-692
3,5-Dibromo-1-methylpyrazin-2(1H)-one 97%
3,5-Dibromo-1-methylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C5H4Br2N2O
MW: 267.9061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12022617
CAS Number: 87486-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR