3,5-Dibromo-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-1,2,4-triazole

A153419-10G A153419-25G A153419-100G A153419-500G
103842-686EA 7 USD
103842-686 77273-052 77318-344 77549-938 77567-224
3,5-Dibromo-1H-1,2,4-triazole 98%
3,5-Dibromo-1H-1,2,4-triazole

Precaución: For research use only.

Formula: C₂HBr₂N₃
MW: 226.8600 g/mol
Boiling Pt: 364.8 °C (760 mm Hg)
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00487260
CAS Number: 7411-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR