3,5-Diaminobenzoic acid 98%

Supplier: Ambeed
Warning

A310521-100G A310521-10G A310521-500G A310521-25G A310521-5G
103835-808EA 5 USD
103835-808 77032-592 77032-594 77032-596 77306-026 77306-028
3,5-Diaminobenzoic acid 98%
3,5-Diaminobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 459.1 °C (760 mm Hg)
Melting Pt: 235…238 °C
Flash Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00007807
CAS Number: 535-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR