1-Ethynyl-3,5-bis(trifluoromethyl)benzene 97%

Supplier: Ambeed
Danger

Synonyms: 3',5'-Bis(trifluoromethyl)phenylacetylene

A248157-1G A248157-25G A248157-5G A248157-10G
103811-816EA 9 USD
103811-816 103851-650 77090-986 77090-988 77090-990 77113-390
1-Ethynyl-3,5-bis(trifluoromethyl)benzene 97%
1-Ethynyl-3,5-bis(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₄F₆
MW: 238.13 g/mol
Boiling Pt: 147…148 °C (760 mm Hg)
Flash Pt: 44.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861850
CAS Number: 88444-81-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR