3,5-Bis(trifluoromethyl)phenol 97%
Supplier: Ambeed
|
Warning
|
A114953-10G
A114953-1G
A114953-25G
A114953-500G
A114953-5G
A114953-100G
103824-346EA
13
USD
103824-346
76766-088
76766-090
76766-092
76766-094
77311-164
77623-962
3,5-Bis(trifluoromethyl)phenol 97%
3,5-Bis(trifluoromethyl)phenol
Precaución: for Research Use Only
Formula:
C₈H₄F₆O Boiling Pt: 176.2 °C (760 mm Hg) Melting Pt: 20…21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000386 CAS Number: 349-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...