3,5-Bis(trifluoromethyl)thiophenol 97%

Supplier: Ambeed
Danger

Synonyms: 3,5-Bis(trifluoromethyl)benzenethiol

A238218-1G A238218-5G A238218-10G
103811-208EA 10 USD
103811-208 103850-992 77090-104 77090-106 77275-942
3,5-Bis(trifluoromethyl)thiophenol 97%
3,5-Bis(trifluoromethyl)thiophenol

Precaución: for Research Use Only

Formula: C₈H₄F₆S
MW: 246.18 g/mol
Boiling Pt: 166.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00042273
CAS Number: 130783-02-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR