3,5-Bis(trifluoromethyl)benzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A111403-10G
A111403-25G
A111403-5G
A111403-100G
A111403-500G
103823-646EA
6
USD
103823-646
76687-598
76687-600
77310-454
77549-162
77623-922
3,5-Bis(trifluoromethyl)benzaldehyde 97%
3,5-Bis(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₄F₆O Boiling Pt: 188.5 °C (760 mm Hg) Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010206 CAS Number: 401-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...