3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Bis(trifluoromethyl)-1,2-diaminobenzene

A241002-10G A241002-250MG
103811-386EA 26 USD
103811-386 103851-192 76771-406 76771-408
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₈H₆F₆N₂
MW: 244.14 g/mol
Boiling Pt: 228.2 °C (760 mm Hg)
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD01631430
CAS Number: 367-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR