3,5-Bis(aminomethyl)benzoic acid dihydrochloride 97%

Supplier: Ambeed
Warning

A422767-1G A422767-100MG A422767-5G
103827-426EA 257 USD
103827-426 76842-102 77168-550 77892-873
3,5-Bis(aminomethyl)benzoic acid dihydrochloride 97%
3,5-Bis(aminomethyl)benzoic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28144945
CAS Number: 185963-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR