3,4-Dimethylthiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A210285-250MG A210285-1G A210285-5G
103817-992EA 119 USD
103817-992 103848-686 76785-668 77303-126 77303-128
3,4-Dimethylthiophene-2-carboxylic acid 98%
3,4-Dimethylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706135
CAS Number: 89639-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR