3,4-Dimethylphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dihydroxy(3,4-dimethylphenyl)borane, (3,4-Dimethylphenyl)metaboric acid, 3,4-Dimethylbenzeneboronic acid
A204724-25G
A204724-5G
A204724-100G
A204724-10G
A204724-1G
A204724-500G
103848-230EA
19
USD
103848-230
77086-410
77086-412
77192-430
77192-432
77192-434
77192-436
3,4-Dimethylphenylboronic acid 97%
3,4-Dimethylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol Boiling Pt: 304.9 °C (760 mm Hg) Melting Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009694 CAS Number: 55499-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...