2,3-Dimethylmaleic anhydride 98%

Supplier: Ambeed
Warning

A407814-25G A407814-5G A407814-100G A407814-10G A407814-500G
103838-204EA 5 USD
103838-204 76841-698 76841-700 77252-188 77252-190 77252-192
2,3-Dimethylmaleic anhydride 98%
2,3-Dimethylmaleic anhydride

Precaución: for Research Use Only

Formula: C₆H₆O₃
Boiling Pt: 223 °C (760 mm Hg)
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005523
CAS Number: 766-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR