3,4-Dimethyl-1H-pyrazole-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A113627-100MG
A113627-250MG
A113627-1G
A113627-5G
103798-258EA
49
USD
103798-258
103824-098
76995-368
76995-370
77297-252
77297-254
3,4-Dimethyl-1H-pyrazole-5-carboxylic acid 97%
3,4-Dimethyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C6H8N2O2 MW: 140.1400 g/mol Melting Pt: 276…277 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD15143212 CAS Number: 89831-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...