3,4-Dimethyl-1H-pyrazole Phosphate 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethyl-1H-pyrazole phosphate

A113340-100G A113340-10G A113340-1G A113340-25G A113340-500G A113340-5G
103824-058EA 6 USD
103824-058 77025-868 77025-870 77025-872 77025-874 77025-876 77297-238
3,4-Dimethyl-1H-pyrazole Phosphate 97%
3,4-Dimethyl-1H-pyrazole Phosphate

Precaución: for Research Use Only

Formula: C₅H₁₁N₂O₄P
MW: 194.13 g/mol
Melting Pt: 167…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842544
CAS Number: 202842-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR