3,4-Dimethyl-1H-pyrazole 96%

Supplier: Ambeed
Danger

A200796-100G A200796-10G A200796-1G A200796-25G A200796-5G
103817-240EA 18 USD
103817-240 103847-874 77030-118 77030-120 77085-938 77085-940 77085-942
3,4-Dimethyl-1H-pyrazole 96%
3,4-Dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₈N₂
MW: 96.133 g/mol
Boiling Pt: 220.3 °C (760 mm Hg)
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00462154
CAS Number: 2820-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR