(3,4-Dimethoxyphenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A127474-1000G
A127474-100G
A127474-10G
A127474-500G
A127474-25G
A127474-5G
76870-582EA
168
USD
76870-582
76870-584
76870-586
76870-588
77298-216
77298-218
(3,4-Dimethoxyphenyl)acetic acid 98%
(3,4-Dimethoxyphenyl)acetic acid
Precaución: for Research Use Only
Formula:
(C₆H₃(OCH₃)₂)CH₂COOH Melting Pt: 96…98 °C Flash Pt: 130.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004335 CAS Number: 93-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...