3,4-Dimethoxy-3-cyclobutene-1,2-dione 98%

Supplier: Ambeed
Danger

Synonyms: 1,2-Dimethoxy-1-cyclobutene-3,4-dione, Dimethyl squarate, Squaric acid dimethylester

A229849-1G A229849-25G A229849-5G A229849-10G
103819-500EA 9 USD
103819-500 103850-402 77089-252 77089-254 77089-256 77112-948
3,4-Dimethoxy-3-cyclobutene-1,2-dione 98%
3,4-Dimethoxy-3-cyclobutene-1,2-dione

Precaución: for Research Use Only

Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 286.1 °C (760 mm Hg)
Melting Pt: 54…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00101316
CAS Number: 5222-73-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR